This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295893_m1
See other NDRG1 GE Assays ›
Gene Symbol:
Ndrg1
Gene Name:
N-myc downstream regulated gene 1
Gene Aliases:
CAP43, CMT4D, DRG1, HMSNL, NMSL, Ndr1, Ndrl, PROXY1, RTP, TDD5
Chromosome Location:
Chr.15: 66929318 - 66969641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295893_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndrg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008681.2 NP_032707.2 5 - 6 480 88
GenBank mRNA AK004548.1 - 5 - 6 480 88
AK098075.1 - 5 - 6 479 88
AK161860.1 - 5 - 6 482 88
AK165007.1 - 5 - 6 480 88
AK170281.1 - 5 - 6 481 88
AK170439.1 - 5 - 6 479 88
AK170636.1 - 5 - 6 479 88
AK170809.1 - 5 - 6 480 88
AW611038.1 - 5 - 6 480 88
BC015282.1 - 5 - 6 435 4237943 88
BC071235.1 - 5 - 6 488 4242075 88
BC141246.1 - 5 - 6 371 9055858 88
BC171968.1 - 5 - 6 371 9054254 88
CA320478.1 - 2 - 3 223 88
U60593.1 - 5 - 6 478 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndrg1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
peripheral nervous system myelin maintenance
mast cell activation
positive regulation of spindle checkpoint
microtubule binding
Rab GTPase binding
gamma-tubulin binding
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0