This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07294206_s1
See other CEBPB GE Assays ›
Gene Symbol:
Cebpb
Gene Name:
CCAAT/enhancer binding protein (C/EBP), beta
Gene Aliases:
C/EBPbeta, CRP2, IL-6DBP, LAP, LIP, NF-IL6, NF-M, Nfil6
Chromosome Location:
Chr.2: 167688915 - 167690432 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07294206_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cebpb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287738.1 NP_001274667.1 1 322 105
NM_001287739.1 NP_001274668.1 1 322 105
NM_009883.4 NP_034013.1 1 322 105
GenBank mRNA AK143374.1 - 1 271 105
M61007.1 - 1 231 105
X62600.1 - 1 322 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001287739.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cebpb

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
embryonic placenta development
granuloma formation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cell differentiation
neuron differentiation
response to lipopolysaccharide
positive regulation of interleukin-4 production
mammary gland epithelial cell proliferation
response to endoplasmic reticulum stress
T-helper 1 cell activation
negative regulation of T cell proliferation
defense response to bacterium
negative regulation of neuron apoptotic process
regulation of interleukin-6 biosynthetic process
fat cell differentiation
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
regulation of osteoclast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
brown fat cell differentiation
mammary gland epithelial cell differentiation
regulation of transcription involved in cell fate commitment
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to amino acid stimulus
hepatocyte proliferation
liver regeneration
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
kinase binding
histone acetyltransferase binding
glucocorticoid receptor binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
ubiquitin-like protein ligase binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0