This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07293093_m1
See other DHRS7B GE Assays ›
Gene Symbol:
Dhrs7b
Gene Name:
dehydrogenase/reductase (SDR family) member 7B
Gene Aliases:
BC003479, C79874, C80074, PexRAP
Chromosome Location:
Chr.11: 60830631 - 60858423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07293093_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs7b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172112.1 NP_001165583.1 2 - 3 313 77
NM_145428.2 NP_663403.1 2 - 3 284 77
GenBank mRNA AK052209.1 - 2 - 3 236 77
AK139084.1 - 2 - 3 265 77
AK144057.1 - 2 - 3 219 77
BC003479.1 - 2 - 3 239 2655855 77
JX196872.1 - 2 - 3 203 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhrs7b

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
oxidation-reduction process
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0