This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05916762_s1
See other QSOX1 GE Assays ›
Gene Symbol:
Qsox1
Gene Name:
quiescin Q6 sulfhydryl oxidase 1
Gene Aliases:
1300003H02Rik, QSOX, Qscn6, SOx
Chromosome Location:
Chr.1: 155778155 - 155812899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05916762_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Qsox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024945.1 NP_001020116.1 12 2767 119
NM_023268.2 NP_075757.1 13 1901 119
GenBank mRNA AK004880.1 - 13 1847 119
AK012943.1 - 3 446 119
AK083938.1 - 12 2718 119
AK166839.1 - 12 2715 119
AK169877.1 - 13 2133 119
AK170449.1 - 13 1814 119
AK171601.1 - 119
BC034131.1 - 10 1393 119
BC076590.1 - 12 2720 30548238 119

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Qsox1

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

protein folding
negative regulation of macroautophagy
cell redox homeostasis
oxidation-reduction process
protein disulfide isomerase activity
oxidoreductase activity
flavin-linked sulfhydryl oxidase activity
thiol oxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0