This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05912455_s1
See other ARHGEF7 GE Assays ›
Gene Symbol:
Arhgef7
Gene Name:
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases:
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location:
Chr.8: 11728105 - 11835219 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05912455_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgef7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113518.1 NP_001106990.1 21 4420 95
NM_017402.4 NP_059098.2 20 4243 95
GenBank mRNA AF247654.1 - 21 4313 95
AF247655.1 - 18 3804 95
AK052545.1 - 20 4214 95
AW046058.1 - 1 95
AY220301.1 - 21 4382 95
BC026704.1 - 1 467 95
BC044838.1 - 20 4228 3499604 95
U96634.1 - 18 3931 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgef7

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
small GTPase mediated signal transduction
nervous system development
positive regulation of fibroblast migration
lamellipodium assembly
positive regulation of protein binding
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
astrocyte cell migration
focal adhesion assembly
positive regulation of growth hormone secretion
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0