This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05910460_s1
See other PPP5C GE Assays ›
Gene Symbol:
Ppp5c
Gene Name:
protein phosphatase 5, catalytic subunit
Gene Aliases:
AU020526, PP5
Chromosome Location:
Chr.7: 17004640 - 17027914 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05910460_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp5c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011155.2 NP_035285.2 1 149 74
GenBank mRNA AF018262.1 - 1 120 74
BB868382.1 - 1 130 74
BC003744.1 - 1 105 3590322 74
BY287105.1 - 1 74
DV653678.1 - 1 86 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp5c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
protein dephosphorylation
signal transduction
dephosphorylation
histone dephosphorylation
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to morphine
protein oligomerization
protein heterooligomerization
negative regulation of cell death
negative regulation of neuron death
positive regulation of glucocorticoid receptor signaling pathway
G-protein alpha-subunit binding
RNA binding
phosphoprotein phosphatase activity
signal transducer activity
protein binding
microtubule binding
hydrolase activity
heat shock protein binding
identical protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0