This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05908945_s1
See other TBC1D1 GE Assays ›
Gene Symbol:
Tbc1d1
Gene Name:
TBC1 domain family, member 1
Gene Aliases:
1110062G02Rik, AI385682, AW555803, Tbc1, mKIAA1108
Chromosome Location:
Chr.5: 64156310 - 64351486 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05908945_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tbc1d1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289514.1 NP_001276443.1 19 4481 89
NM_019636.3 NP_062610.2 20 5062 89
GenBank mRNA AK004348.1 - 1 661 89
AK122445.1 - 22 6150 89
AK148356.1 - 19 4438 89
AK154591.1 - 21 5097 89
AK168763.1 - 10 2732 89
BC004675.1 - 9 2720 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tbc1d1

Panther Classification:

Molecular Function -

scaffold/adaptor protein GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein localization
GTPase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0