This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05908665_s1
See other VDR GE Assays ›
Gene Symbol:
Vdr
Gene Name:
vitamin D receptor
Gene Aliases:
Nr1i1
Chromosome Location:
Chr.15: 97854427 - 97908296 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05908665_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vdr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009504.4 NP_033530.2 10 3433 115
GenBank mRNA AK030339.1 - 10 3435 115
AK156789.1 - 10 3437 115

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vdr

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
skeletal system development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
calcium ion transport
cellular calcium ion homeostasis
multicellular organism development
lactation
negative regulation of cell proliferation
organ morphogenesis
positive regulation of gene expression
negative regulation of keratinocyte proliferation
positive regulation of vitamin D 24-hydroxylase activity
intracellular receptor signaling pathway
bile acid signaling pathway
steroid hormone mediated signaling pathway
positive regulation of keratinocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
intestinal absorption
regulation of calcium ion transport
positive regulation of apoptotic process involved in mammary gland involution
regulation of calcidiol 1-monooxygenase activity
mammary gland branching involved in pregnancy
vitamin D receptor signaling pathway
apoptotic signaling pathway
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
vitamin D binding
protein binding
zinc ion binding
calcitriol receptor activity
lithocholic acid receptor activity
sequence-specific DNA binding
metal ion binding
retinoid X receptor binding
vitamin D response element binding
calcitriol binding
lithocholic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0