This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05907543_s1
See other RCAN3 GE Assays ›
Gene Symbol:
Rcan3
Gene Name:
regulator of calcineurin 3
Gene Aliases:
AU041093, Csp3, Dscr1l2
Chromosome Location:
Chr.4: 135412309 - 135433805 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05907543_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rcan3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022980.4 NP_075356.1 5 4865 92
GenBank mRNA AK155270.1 - 5 4866 92
BC059001.1 - 4 4647 6847734 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rcan3

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

calcium-mediated signaling
regulation of catalytic activity
regulation of calcineurin-NFAT signaling cascade
nucleotide binding
calcium-dependent protein serine/threonine phosphatase regulator activity
troponin I binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header