This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05906664_s1
See other PRKCI GE Assays ›
Gene Symbol:
Prkci
Gene Name:
protein kinase C, iota
Gene Aliases:
2310021H13Rik, AI427505, PKClambda, Pkci, Pkcl, Prkcl, aPKClambda, mKIAA4165
Chromosome Location:
Chr.3: 30995742 - 31052741 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05906664_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkci
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008857.3 NP_032883.2 18 4192 60
GenBank mRNA AK009442.1 - 1 1895 60
AK144979.1 - 18 4188 60
AK167411.1 - 18 4161 60
AK220517.1 - 18 4216 60
BC021630.1 - 18 4063 3980628 60
BC023253.1 - 1 1423 60

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkci

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
actin filament organization
multicellular organism development
positive regulation of neuron projection development
phosphorylation
cell migration
cellular response to insulin stimulus
negative regulation of glial cell apoptotic process
cellular protein localization
establishment of apical/basal cell polarity
intracellular signal transduction
eye photoreceptor cell development
response to peptide hormone
negative regulation of neuron apoptotic process
cell-cell junction organization
positive regulation of glucose import
Golgi vesicle budding
positive regulation of NF-kappaB transcription factor activity
positive regulation of glial cell proliferation
response to interleukin-1
positive regulation of establishment of protein localization to plasma membrane
positive regulation of endothelial cell apoptotic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein kinase C activity
ATP binding
phospholipid binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein domain specific binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0