This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05836962_gH
See other RPLP0 GE Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
36B4, Arbp, L10E
Chromosome Location:
Chr.5: 115559467 - 115563729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05836962_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rplp0
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007475.5 NP_031501.1 6 844 83
GenBank mRNA AK002315.1 - 6 797 83
AK009767.1 - 6 798 84
AK010267.1 - 6 797 83
AK012606.1 - 6 803 83
AK029816.1 - 6 799 83
AK081678.1 - 6 801 83
AK150448.1 - 6 797 83
AK159358.1 - 6 797 83
AK160541.1 - 6 797 83
AK161683.1 - 6 799 83
AK166728.1 - 6 797 83
AK167240.1 - 6 795 83
BC003833.1 - 6 787 3600590 83
BC011106.1 - 5 772 3982438 83
BC011291.1 - 5 778 3588342 83
BC030189.1 - 4 584 83
BC057295.1 - 5 778 83
BC087887.1 - 6 798 6594755 83
BC089496.1 - 6 798 30254142 83
BC099384.1 - 5 764 30843295 83
X15267.1 - 6 796 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rplp0

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

ribosome biogenesis
cellular response to interleukin-4
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0