This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05829898_s1
See other RGS2 GE Assays ›
Gene Symbol:
Rgs2
Gene Name:
regulator of G-protein signaling 2
Gene Aliases:
GOS8
Chromosome Location:
Chr.1: 143999338 - 144004149 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05829898_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009061.4 NP_033087.2 5 1395 96
GenBank mRNA AK029310.1 - 5 1396 96
AK031603.1 - 5 1398 96
AK077922.1 - 5 1396 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs2

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
cell cycle
G-protein coupled receptor signaling pathway
negative regulation of signal transduction
negative regulation of phospholipase activity
negative regulation of cardiac muscle hypertrophy
positive regulation of microtubule polymerization
termination of G-protein coupled receptor signaling pathway
negative regulation of MAP kinase activity
positive regulation of GTPase activity
negative regulation of cAMP-mediated signaling
negative regulation of G-protein coupled receptor protein signaling pathway
brown fat cell differentiation
relaxation of cardiac muscle
relaxation of vascular smooth muscle
positive regulation of cardiac muscle contraction
regulation of adrenergic receptor signaling pathway
GTPase activator activity
protein binding
beta-tubulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header