This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05776793_s1
See other SEMA3B GE Assays ›
Gene Symbol:
Sema3b
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B
Gene Aliases:
SemA, Semaa, sema5, semaV
Chromosome Location:
Chr.9: 107597674 - 107609303 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05776793_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042779.2 NP_001036244.1 20 2746 91
NM_001291537.1 NP_001278466.1 20 3010 91
NM_001291538.1 NP_001278467.1 20 3004 91
NM_001291539.1 NP_001278468.1 19 2687 91
NM_009153.3 NP_033179.2 18 2556 91
NR_111986.1 18 2499 91
GenBank mRNA AK014333.1 - 18 2435 91
AK033597.1 - 19 2697 91
AK037034.1 - 20 2735 91
AK043634.1 - 19 2640 91
AK048364.1 - 19 2968 91
AK148232.1 - 20 2708 91
AK195512.1 - 1 118 91
AK198160.1 - 1 118 91
AK204724.1 - 1 118 91
AK207568.1 - 1 118 91
AK218182.1 - 1 118 91
BC090669.1 - 19 2667 30696359 91
BC150758.1 - 17 2402 9087669 91
X85990.1 - 18 2553 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema3b

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s)

multicellular organism development
nervous system development
cell differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0