This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05679361_s1
See other PIK3CA GE Assays ›
Gene Symbol:
Pik3ca
Gene Name:
phosphatidylinositol 3-kinase, catalytic, alpha polypeptide
Gene Aliases:
6330412C24Rik, caPI3K, p110, p110alpha
Chromosome Location:
Chr.3: 32397055 - 32468486 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05679361_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pik3ca
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008839.2 NP_032865.2 20 85
GenBank mRNA AK053528.1 - 1 85
AK078608.1 - 1 423 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pik3ca

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
liver development
adaptive immune response
glucose metabolic process
protein phosphorylation
inflammatory response
phosphatidylinositol 3-kinase signaling
phosphorylation
platelet activation
positive regulation of peptidyl-serine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
regulation of cellular respiration
protein kinase B signaling
negative regulation of neuron apoptotic process
hypomethylation of CpG island
innate immune response
positive regulation of protein kinase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
cell chemotaxis
adipose tissue development
cellular response to glucose stimulus
energy homeostasis
mitophagy in response to mitochondrial depolarization
negative regulation of fibroblast apoptotic process
regulation of genetic imprinting
negative regulation of anoikis
nucleotide binding
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
1-phosphatidylinositol-3-kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphotransferase activity, alcohol group as acceptor
protein kinase activator activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0