This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934720_s1
See other DALIR GE Assays ›
Gene Symbol:
Dalir
Gene Name:
DNMT1 associated long intergenic non-coding RNA
Gene Aliases:
Dali, Gm28525
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934720_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dalir
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK034039.1 - 1 2016 103
AW489559.1 - 1 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB078011.2 - 1 507 Chr. 1 - 42752221 - 42752873 [+] on Build GRCm38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream POU domain,
class 3,
transcription factor 3 adjacent noncoding transcript 2
Pantr2 2610207O16Rik,
Pou3f3os2,
linc-Brn1b
-38.8kb Anti-sense Chr. 1 - 42707061 - 42713454 on NCBI Build GRCm38
Downstream mitochondrial ribosomal protein S9 Mrps9 2310002A08Rik,
AW322516,
Mrp-S9,
S9mt,
mg637
98.4kb Sense Chr. 1 - 42851233 - 42905683 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0