This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934650_m1
See other 4930596I21RIK GE Assays ›
Gene Symbol:
4930596I21Rik
Gene Name:
RIKEN cDNA 4930596I21 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 138977657 - 138979246 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934650_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930596I21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_108103.1 2 - 3 371 101
GenBank mRNA AK019844.1 - 2 - 3 372 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY716049.1 - 2 - 3 372 Chr. 1 - 138977657 - 138979254 [+] on Build GRCm38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 1700019P21 gene 1700019P21Rik -101.1kb Anti-sense Chr. 1 - 138876272 - 138876965 on NCBI Build GRCm38
Downstream crumbs family member 1,
photoreceptor morphogenesis associated
Crb1 7530426H14Rik,
A930008G09Rik
218.3kb Anti-sense Chr. 1 - 139197546 - 139379118 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0