This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934607_s1
See other MIR7016 GE Assays ›
Gene Symbol:
Mir7016
Gene Name:
microRNA 7016
Gene Aliases:
Gm27994, mmu-mir-7016
Chromosome Location:
Chr.4: 129684500 - 129684571 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934607_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir7016
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105983.1 1 43 55

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane protein 39b Tmem39b 6330509E05Rik,
BC037095
Sense Chr. 4 - 129676355 - 129696851 on NCBI Build GRCm38
Downstream KH domain containing,
RNA binding,
signal transduction associated 1
Khdrbs1 Sam68,
p62,
p68
18.6kb Sense Chr. 4 - 129703164 - 129742768 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0