This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04416583_s1
See other MIR1198 GE Assays ›
Gene Symbol:
Mir1198
Gene Name:
microRNA 1198
Gene Aliases:
Mirn1198, mmu-mir-1198
Chromosome Location:
Chr.X: 7807101 - 7807221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04416583_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1198
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035430.1 1 61 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream GRIP1 associated protein 1 Gripap1 AI854681,
DXImx47e,
GRASP-1,
Sfc10,
mKIAA1167
Sense Chr. X - 7789961 - 7820567 on NCBI Build GRCm38
Downstream potassium voltage-gated channel,
Shal-related family,
member 1
Kcnd1 1110037K09Rik,
Kca2-1,
Kv4.1,
Shal,
Shal1,
mShal1
16.6kb Sense Chr. X - 7823843 - 7836503 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to inorganic substance

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0