This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04416442_s1
See other MIR6539 GE Assays ›
Gene Symbol:
Mir6539
Gene Name:
microRNA 6539
Gene Aliases:
Gm27464, mir-6539, mmu-mir-6539
Chromosome Location:
Chr.14: 67859636 - 67859745 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04416442_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6539
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105857.1 1 63 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream gonadotropin releasing hormone 1 Gnrh1 Gnrh,
Gnrh2,
LHRH,
Lhrh1,
Lnrh,
hpg
-110.2kb Anti-sense Chr. 14 - 67745229 - 67749446 on NCBI Build GRCm38
Downstream neurofilament,
light polypeptide
Nefl AI847934,
CMT2E,
NF-L,
NF68,
Nfl
224.1kb Anti-sense Chr. 14 - 68083884 - 68087745 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017768.mm.1 NR_105857.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017768.mm.1 TC1400002294.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0