This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04416440_s1
See other MIR6416 GE Assays ›
Gene Symbol:
Mir6416
Gene Name:
microRNA 6416
Gene Aliases:
Gm27746, mmu-mir-6416
Chromosome Location:
Chr.7: 64249983 - 64250099 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04416440_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6416
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105844.1 1 43 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transient receptor potential cation channel,
subfamily M,
member 1
Trpm1 4732499L03Rik,
AI606771,
Ltrpc1,
Mlsn1
Anti-sense Chr. 7 - 64153835 - 64269772 on NCBI Build GRCm38
Downstream myotubularin related protein 10 Mtmr10 BB128963,
C79734
37.6kb Anti-sense Chr. 7 - 64287670 - 64340806 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700029082.mm.1 NR_105844.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029082.mm.1 TC0700003409.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6889b944d9-flx6z:80/100.66.128.142:80.
git-commit: df865406a845caddbd23a531a4b79e504db49c75
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header