This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411729_m1
See other ZEB2OS GE Assays ›
Gene Symbol:
Zeb2os
Gene Name:
zinc finger E-box binding homeobox 2, opposite strand
Gene Aliases:
Gm13476, OTTMUSG00000012358
Chromosome Location:
Chr.2: 45111138 - 45114087 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411729_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zeb2os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110572.1 1 - 2 163 76
GenBank mRNA BC031611.1 - 1 - 2 163 76
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB204594.1 - 82 Chr. - - [-] on Build 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 5129 Mir5129 mir-5129,
mmu-mir-5129,
mu-mir-5129
-88.0kb Anti-sense Chr. 2 - 45023100 - 45023177 on NCBI Build GRCm38
Downstream viral integration site 1 Vis1 Gm27276,
His-1,
His1
126.0kb Sense Chr. 2 - 45238980 - 45282900 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0