This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411611_m1
See other CYP2R1 GE Assays ›
Gene Symbol:
Cyp2r1
Gene Name:
cytochrome P450, family 2, subfamily r, polypeptide 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 114550123 - 114565618 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411611_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cyp2r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110408.1 1 - 2 234 99
GenBank mRNA AK050453.1 - 1 - 2 224 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB654100.1 - 1 - 2 220 Chr. 7 - 114551761 - 114562958 [-] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphodiesterase 3B,
cGMP-inhibited
Pde3b 9830102A01Rik,
AI847709
-16.7kb Anti-sense Chr. 7 - 114414700 - 114537938 on NCBI Build GRCm38
Downstream calcitonin/calcitonin-related polypeptide,
alpha
Calca CA,
CGRP-1,
CGRP1,
Calc,
Calc1,
Cgrp,
Ct,
Ctn
63.0kb Sense Chr. 7 - 114625985 - 114636460 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK050453.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

organic acid metabolic process
xenobiotic metabolic process
calcitriol biosynthetic process from calciol
vitamin D metabolic process
exogenous drug catabolic process
oxidation-reduction process
monooxygenase activity
iron ion binding
steroid hydroxylase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
oxygen binding
heme binding
vitamin D3 25-hydroxylase activity
metal ion binding
cholestanetriol 26-monooxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0