This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411524_m1
See other ST6GALNAC6 GE Assays ›
Gene Symbol:
St6galnac6
Gene Name:
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases:
ST6GalNAcVI, Siat7f
Chromosome Location:
Chr.2: 32599709 - 32620809 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411524_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: St6galnac6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110347.1 5 - 6 839 60
GenBank mRNA BC022924.1 - 5 - 6 783 4208472 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 St6galnac4 SIAT7-D,
ST6GalNAcIV,
Siat7d
-15.1kb Sense Chr. 2 - 32587078 - 32599696 on NCBI Build GRCm38
Downstream adenylate kinase 1 Ak1 Ak-1,
B430205N08Rik
3.2kb Sense Chr. 2 - 32621758 - 32635058 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000587.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ganglioside biosynthetic process
protein glycosylation
glycosylceramide metabolic process
glycosphingolipid metabolic process
glycoprotein metabolic process
oligosaccharide biosynthetic process
cell-cell recognition
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
lytic endotransglycosylase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NM_001025310.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NM_001025311.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NM_001289547.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NM_001289548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NM_001289549.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NM_016973.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 NR_110347.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005357.mm.1 TC0200000587.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NM_001025310.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NM_001025311.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NM_001289547.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NM_001289548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NM_001289549.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NM_016973.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 NR_110347.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005360.mm.1 TC0200000587.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0