This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411482_m1
See other DGKEOS GE Assays ›
Gene Symbol:
Dgkeos
Gene Name:
diacylglycerol kinase, epsilon, opposite strand
Gene Aliases:
A930013B10Rik
Chromosome Location:
Chr.11: 89060751 - 89067884 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411482_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgkeos
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110336.1 1 - 2 76 74
GenBank mRNA BY080538.1 - 1 - 2 76 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream diacylglycerol kinase,
epsilon
Dgke C87606,
DAGK6,
DGK
Anti-sense Chr. 11 - 89037582 - 89060763 on NCBI Build GRCm38
Downstream predicted gene 525 Gm525 7.6kb Sense Chr. 11 - 89073841 - 89093064 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001388.mm.1
This assay targets the same location as RTA transcript cluster(s) TC1000001375.rn

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100013014.mm.1 NR_110336.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100013014.mm.1 TC1100001388.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100013015.mm.1 NR_110336.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100013015.mm.1 TC1100001388.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100013010.mm.1 NR_110336.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100013010.mm.1 TC1100001388.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0