This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411420_m1
See other 1700109K24RIK GE Assays ›
Gene Symbol:
1700109K24Rik
Gene Name:
RIKEN cDNA 1700109K24 gene
Gene Aliases:
4933425G21Rik
Chromosome Location:
Chr.15: 77084398 - 77096314 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411420_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700109K24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_108037.1 2 - 3 623 93
GenBank mRNA AK007154.1 - 2 - 3 578 93
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY707238.1 - 2 - 3 577 Chr. 15 - 77084443 - 77096279 [+] on Build GRCm38 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RNA binding protein,
fox-1 homolog (C. elegans) 2
Rbfox2 Fbm2,
Fxh,
Hrnbp2,
Rbm9
Anti-sense Chr. 15 - 77078990 - 77307053 on NCBI Build GRCm38
Downstream uncharacterized LOC102639762 LOC102639762 273.2kb Anti-sense Chr. 15 - 77369492 - 77377210 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0