This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411403_s1
See other MIR8105 GE Assays ›
Gene Symbol:
Mir8105
Gene Name:
microRNA 8105
Gene Aliases:
Gm27346, mmu-mir-8105
Chromosome Location:
Chr.10: 128458680 - 128458768 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411403_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir8105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_106183.1 1 47 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ring finger protein 41 Rnf41 2210404G21Rik,
4930511A05Rik,
4933415P08Rik,
D10Ertd722e,
FLRF,
Nrdp1
-17.2kb Sense Chr. 10 - 128411616 - 128441441 on NCBI Build GRCm38
Downstream SWI/SNF related,
matrix associated,
actin dependent regulator of chromatin,
subfamily c,
member 2
Smarcc2 5930405J04Rik 468.0 Sense Chr. 10 - 128459236 - 128490583 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000011821.mm.1 NR_106183.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011821.mm.1 TC1000001608.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0