This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411342_s1
See other MIR6769B GE Assays ›
Gene Symbol:
Mir6769b
Gene Name:
microRNA 6769b
Gene Aliases:
Gm27546, mmu-mir-6769b
Chromosome Location:
Chr.8: 71631047 - 71631106 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411342_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6769b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_106035.1 1 36 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream collagen beta(1-O)galactosyltransferase 1 Colgalt1 2810024B22Rik,
Glt25d1
-6.1kb Anti-sense Chr. 8 - 71610991 - 71624911 on NCBI Build GRCm38
Downstream Janus kinase 3 Jak3 fae,
wil
45.3kb Anti-sense Chr. 8 - 71676383 - 71690577 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800018656.mm.1 NR_106035.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800018656.mm.1 TC0800002438.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0