This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411331_s1
See other MIR7034 GE Assays ›
Gene Symbol:
Mir7034
Gene Name:
microRNA 7034
Gene Aliases:
Gm27372, mmu-mir-7034
Chromosome Location:
Chr.5: 135735806 - 135735889 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411331_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir7034
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_106001.1 1 45 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream P450 (cytochrome) oxidoreductase Por 4933424M13Rik,
CPR,
CYPOR,
P450R
-480.0 Anti-sense Chr. 5 - 135670040 - 135735326 on NCBI Build GRCm38
Downstream serine/threonine/tyrosine interacting-like 1 Styxl1 1700011C14Rik,
Dusp24,
Mkstyx
11.3kb Sense Chr. 5 - 135747220 - 135778410 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030766.mm.1 NR_106001.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030766.mm.1 TC0500003405.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0