This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411328_s1
See other MIR7025 GE Assays ›
Gene Symbol:
Mir7025
Gene Name:
microRNA 7025
Gene Aliases:
Gm27860, mmu-mir-7025
Chromosome Location:
Chr.5: 75176831 - 75176904 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411328_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir7025
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105992.1 1 40 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
19583
Gm19583 -29.7kb Sense Chr. 5 - 75110321 - 75147107 on NCBI Build GRCm38
Downstream kit oncogene Kit Bs,
CD117,
Fdc,
Gsfsco1,
Gsfsco5,
Gsfsow3,
SCO1,
SCO5,
SOW3,
Ssm,
Tr-kit,
W,
c-KIT
398.1kb Sense Chr. 5 - 75574987 - 75656722 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500006262.mm.1 NR_105992.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500006262.mm.1 TC0500000709.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0