This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411301_s1
See other MIR6948 GE Assays ›
Gene Symbol:
Mir6948
Gene Name:
microRNA 6948
Gene Aliases:
mmu-mir-6948
Chromosome Location:
Chr.14: 54643049 - 54643110 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411301_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6948
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105913.1 1 33 62
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DT912842.1 - 1 Chr. 14 - 54642753 - 54643359 [+] on Build GRCm38 62
GenBankmRNA AW743940.1 - 1 110 Chr. 14 - 54642726 - 54643187 [-] on Build GRCm38 62

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream apoptotic chromatin condensation inducer 1 Acin1 2610036I19Rik,
2610510L13Rik,
Acinus,
Acn,
C79325,
acinusL,
acinusS,
mKIAA0670
Sense Chr. 14 - 54642161 - 54686931 on NCBI Build GRCm38
Downstream RIKEN cDNA 4930579G18 gene 4930579G18Rik 7.4kb Anti-sense Chr. 14 - 54650511 - 54655100 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015597.mm.1 NR_105913.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015597.mm.1 TC1400002064.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0