This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411298_s1
See other MIR6942 GE Assays ›
Gene Symbol:
Mir6942
Gene Name:
microRNA 6942
Gene Aliases:
Gm27287, mmu-mir-6942
Chromosome Location:
Chr.13: 21396578 - 21396638 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411298_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6942
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105907.1 1 26 61
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ560764.1 - 1 125 Chr. 13 - 21396468 - 21396737 [-] on Build GRCm38 61
GenBankmRNA CA564672.1 - 1 125 Chr. 13 - 21396303 - 21396737 [-] on Build GRCm38 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger and SCAN domain containing 12 Zscan12 2510038J07Rik,
FPM315,
Zfp96,
mKIAA0426
-24.3kb Anti-sense Chr. 13 - 21362820 - 21372302 on NCBI Build GRCm38
Downstream piggyBac transposable element derived 1 Pgbd1 4921509E05Rik 24.6kb Sense Chr. 13 - 21421275 - 21441291 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300009746.mm.1 NR_105907.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300009746.mm.1 TC1300001538.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0