This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411287_s1
See other MIR6913 GE Assays ›
Gene Symbol:
Mir6913
Gene Name:
microRNA 6913
Gene Aliases:
Gm27788, mmu-mir-6913
Chromosome Location:
Chr.10: 81386347 - 81386412 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411287_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6913
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105878.1 1 29 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA930009.1 - 1 196 Chr. 10 - 81386179 - 81387462 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream fizzy/cell division cycle 20 related 1 (Drosophila) Fzr1 AW108046,
Cdh1,
FZR,
FZR2,
Fyr,
HCDH,
HCDH1
-8.0kb Anti-sense Chr. 10 - 81366879 - 81378370 on NCBI Build GRCm38
Downstream small integral membrane protein 24 Smim24 2210404O07Rik 6.7kb Sense Chr. 10 - 81393064 - 81395079 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000007102.mm.1 NR_105878.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007102.mm.1 TC1000000897.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0