This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411281_s1
See other MIR6901 GE Assays ›
Gene Symbol:
Mir6901
Gene Name:
microRNA 6901
Gene Aliases:
Gm27732, mmu-mir-6901
Chromosome Location:
Chr.1: 93274549 - 93274610 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411281_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6901
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105866.1 1 31 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sushi,
nidogen and EGF-like domains 1
Sned1 6720455I24Rik,
AI197264,
AI642697,
D430044C15Rik,
SST3,
Snep
Sense Chr. 1 - 93235815 - 93301065 on NCBI Build GRCm38
Downstream mitochondrial transcription termination factor 4 Mterf4 1810059A23Rik,
4933412H03Rik,
Mterfd2
24.6kb Anti-sense Chr. 1 - 93299211 - 93305965 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100007853.mm.1 NR_105866.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007853.mm.1 TC0100000927.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0