This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411268_s1
See other MIR6417 GE Assays ›
Gene Symbol:
Mir6417
Gene Name:
microRNA 6417
Gene Aliases:
Gm27334, mmu-mir-6417
Chromosome Location:
Chr.5: 52135902 - 52136017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411268_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6417
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105845.1 1 60 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA139737.1 - 1 195 Chr. 5 - 52135772 - 52136152 [-] on Build GRCm38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream peroxisome proliferative activated receptor,
gamma,
coactivator 1 alpha
Ppargc1a A830037N07Rik,
ENSMUSG00000079510,
Gm11133,
PGC-1,
PGC-1v,
Pgc-1alpha,
Pgc1,
Pgco1,
Ppargc1
-19.9kb Sense Chr. 5 - 51454249 - 52115958 on NCBI Build GRCm38
Downstream DEAH (Asp-Glu-Ala-His) box polypeptide 15 Dhx15 DBP1,
DEAH9,
Ddx15,
HRH2,
mDEAH9
14.2kb Sense Chr. 5 - 52150203 - 52190546 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500021697.mm.1 NR_105845.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021697.mm.1 TC0500002404.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0