This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411253_s1
See other MIR378C GE Assays ›
Gene Symbol:
Mir378c
Gene Name:
microRNA 378c
Gene Aliases:
Gm27930, mmu-mir-378c
Chromosome Location:
Chr.14: 46954830 - 46954928 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411253_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir378c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105813.1 1 55 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sterile alpha motif domain containing 4 Samd4 1700024G08Rik,
1700111L17Rik,
4933436G17Rik,
Samd4a,
Smaug,
Smaug1
Anti-sense Chr. 14 - 46882965 - 47105817 on NCBI Build GRCm38
Downstream predicted gene 10371 Gm10371 ENSMUSG00000072589 142.8kb Sense Chr. 14 - 47097679 - 47116161 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300007783.mm.1 NR_105813.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300007783.mm.1 TC0300000963.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header