This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411233_s1
See other MIR6359 GE Assays ›
Gene Symbol:
Mir6359
Gene Name:
microRNA 6359
Gene Aliases:
Gm27328, mmu-mir-6359
Chromosome Location:
Chr.18: 88972030 - 88972110 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411233_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir6359
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105777.1 1 45 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream rotatin Rttn 4921538A15Rik,
AI666264,
C530033I08Rik,
D230040K24
Sense Chr. 18 - 88971738 - 89131016 on NCBI Build GRCm38
Downstream CD226 antigen Cd226 BC051526,
DNAM-1,
DNAM1,
Pta1,
TLiSA1
204.8kb Sense Chr. 18 - 89176954 - 89272232 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800006537.mm.1 NM_175542.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006537.mm.1 NM_175542.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800006537.mm.1 NR_105777.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006537.mm.1 TC1800000908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800006538.mm.1 NR_105777.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006538.mm.1 TC1800000908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header