This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410197_m1
See other CREBZF GE Assays ›
Gene Symbol:
Crebzf
Gene Name:
CREB/ATF bZIP transcription factor
Gene Aliases:
1110034C16Rik, 6330417B10Rik, LAZip, LAZipII, SMILE, Zf
Chromosome Location:
Chr.7: 90442735 - 90448043 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410197_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crebzf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073438.1 3 - 4 1448 90
GenBank mRNA AA028814.1 - 1 - 2 90
CF616479.1 - 1 - 2 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV327114.2 - 3 - 4 465 Chr. 7 - 90443900 - 90447713 [+] on Build GRCm38 90
GenBankmRNA AW913497.1 - 2 - 3 136 Chr. 7 - 90444305 - 90447554 [+] on Build GRCm38 90
GenBankmRNA AA444626.1 - 1 - 2 64 Chr. 7 - 90445424 - 90447595 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 89 Ccdc89 1700019B01Rik,
AU022100,
BOIP
-16.8kb Sense Chr. 7 - 90426312 - 90428664 on NCBI Build GRCm38
Downstream transmembrane protein 126A Tmem126a 1810020E01Rik 2.7kb Anti-sense Chr. 7 - 90450712 - 90457208 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
response to virus
negative regulation of transcription, DNA-templated
regulation of sequence-specific DNA binding transcription factor activity
transcription factor activity, sequence-specific DNA binding
protein binding
identical protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0