This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410095_m1
See other 4932415M13RIK GE Assays ›
Gene Symbol:
4932415M13Rik
Gene Name:
RIKEN cDNA 4932415M13 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 53716509 - 53733843 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410095_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4932415M13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073205.1 6 - 7 2385 63
GenBank mRNA AK030017.1 - 6 - 7 2386 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream shugoshin-like 1 (S. pombe) Sgol1 3300001M08Rik,
C81037,
Sgo1
-39.7kb Anti-sense Chr. 17 - 53674786 - 53689846 on NCBI Build GRCm38
Downstream sulfotransferase family,
cytosolic,
1C,
member 2
Sult1c2 1810008N17Rik,
ST1C1,
ST1C2,
Sult1c1
98.2kb Anti-sense Chr. 17 - 53829637 - 53845958 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000978.mm.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
biological_process
intracellular signal transduction
molecular_function
protein serine/threonine kinase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009362.mm.1 NR_073205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009362.mm.1 TC1700000978.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009368.mm.1 NR_073205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009368.mm.1 TC1700000978.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009369.mm.1 NR_073205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009369.mm.1 TC1700000978.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009364.mm.1 NR_073205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009364.mm.1 TC1700000978.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009366.mm.1 NR_073205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009366.mm.1 TC1700000978.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 750558 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0