This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410089_s1
See other MIR5617 GE Assays ›
Gene Symbol:
Mir5617
Gene Name:
microRNA 5617
Gene Aliases:
Gm25192, mir-5617, mmu-mir-5617
Chromosome Location:
Chr.X: 20863126 - 20863182 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410089_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir5617
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_049190.1 1 27 52

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream synapsin I Syn1 Syn-1,
Syn1-S
Sense Chr. X - 20860511 - 20920918 on NCBI Build GRCm38
Downstream tissue inhibitor of metalloproteinase 1 Timp1 Clgi,
EPA,
TIMP-1,
TPA-S1,
Timp
7.0kb Anti-sense Chr. X - 20870166 - 20874737 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00012259.mm.1 NR_049190.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012259.mm.1 TC0X00001932.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header