This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335964_s1
See other C130030K03RIK GE Assays ›
Gene Symbol:
C130030K03Rik
Gene Name:
proteasome (prosome, macropain) subunit, alpha type 6 pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.10: 49093681 - 49095843 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335964_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C130030K03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046212.1 1 2025 120
GenBank mRNA AK046374.1 - 1 2026 120
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB085994.1 - 1 183 Chr. 10 - 49093681 - 49094000 [-] on Build GRCm38 120
GenBankmRNA BB365727.2 - 1 224 Chr. 10 - 49093335 - 49094042 [-] on Build GRCm38 120

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glutamate receptor,
ionotropic,
kainate 2 (beta 2)
Grik2 AW124492,
GluK2,
Glur-6,
Glur6,
Glurbeta2
Sense Chr. 10 - 49094842 - 49789250 on NCBI Build GRCm38
Downstream activating signal cointegrator 1 complex subunit 3 Ascc3 ASC1p200,
B630009I04Rik,
BC023451,
D430001L07Rik,
D630041L21,
Helic1,
RNAH
1.5mb Anti-sense Chr. 10 - 50592669 - 50851204 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000014974.mm.1 NR_046212.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014974.mm.1 TC1000002120.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014975.mm.1 NR_046212.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014975.mm.1 TC1000002120.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 445311 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0