This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335940_m1
See other 9530052E02RIK GE Assays ›
Gene Symbol:
9530052E02Rik
Gene Name:
RIKEN cDNA 9530052E02 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 11007850 - 11054541 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335940_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9530052E02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046017.1 1 - 2 160 104
GenBank mRNA AK162519.1 - 1 - 2 161 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY722155.1 - 1 - 2 155 Chr. 8 - 11007855 - 11054208 [+] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 3930402G23 gene 3930402G23Rik -79.4kb Anti-sense Chr. 8 - 10924427 - 10928457 on NCBI Build GRCm38
Downstream collagen,
type IV,
alpha 1
Col4a1 Bru,
Col4a-1,
Del(8)44H,
Raw,
Svc
153.6kb Anti-sense Chr. 8 - 11198423 - 11312826 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000084.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800000678.mm.1 NR_046017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000678.mm.1 NR_046017.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000678.mm.1 TC0800000084.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800000677.mm.1 NR_046017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000677.mm.1 NR_046017.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000677.mm.1 TC0800000084.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0