This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335905_m1
See other DNMT3AOS GE Assays ›
Gene Symbol:
Dnmt3aos
Gene Name:
DNA methyltransferase 3A, opposite strand
Gene Aliases:
ENSMUSG00000073242, Gm10485
Chromosome Location:
Chr.12: 3859293 - 3862244 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335905_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnmt3aos
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045884.1 3 - 4 478 55
GenBank mRNA AK150533.1 - 3 - 4 479 55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY743475.1 - 3 - 4 477 Chr. 12 - 3860041 - 3862243 [-] on Build GRCm38 55

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream dystrobrevin,
beta
Dtnb -76.7kb Anti-sense Chr. 12 - 3572391 - 3782611 on NCBI Build GRCm38
Downstream pro-opiomelanocortin-alpha Pomc ACTH,
BE,
Beta-LPH,
Clip,
Gamma-LPH,
Npp,
Pomc-1,
Pomc1,
alpha-MSH,
alphaMSH,
beta-MSH,
gamma-MSH
93.3kb Anti-sense Chr. 12 - 3954945 - 3960643 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK150533.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171785 AK150533.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171785 AK150533.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200009647.mm.1 NR_045884.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009647.mm.1 NR_045884.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009647.mm.1 TC1200001347.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0