This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335204_s1
See other MIRLET7D GE Assays ›
Gene Symbol:
Mirlet7d
Gene Name:
microRNA let7d
Gene Aliases:
Mirnlet7d, let-7d, mmu-let-7d
Chromosome Location:
Chr.13: 48536012 - 48536114 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335204_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mirlet7d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029656.1 1 52 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK160312.1 - 1057 Chr. - - [-] on Build 75
GenBankmRNA BQ031149.1 - 1 Chr. 13 - 48535523 - 48536162 [+] on Build GRCm38 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA A830005F24 gene A830005F24Rik -21.0kb Anti-sense Chr. 13 - 48513540 - 48515002 on NCBI Build GRCm38
Downstream microRNA let7f-1 Mirlet7f-1 Mirlet7f1,
Mirnlet7f1,
let-7f-1
1.7kb Sense Chr. 13 - 48537829 - 48537917 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

embryo implantation
BMP signaling pathway
response to oxygen levels
cellular response to amino acid stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1436540_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10409059 - -
Affymetrix Mouse Clariom S Array PSR1300012091.mm.1 NR_029656.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012091.mm.1 NR_029656.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012091.mm.1 TC1300001941.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0