This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335189_s1
See other MIR1A-1 GE Assays ›
Gene Symbol:
Mir1a-1
Gene Name:
microRNA 1a-1
Gene Aliases:
Mir1-1, Mirn1-1, Mirn1b, Mirn1c, Mirn1d, mir-1a-1, mmu-mir-1-1, mmu-mir-1a-1
Chromosome Location:
Chr.2: 180389048 - 180389124 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335189_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1a-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029528.1 1 44 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 6307 Gm6307 EG622283 Sense Chr. 2 - 180385604 - 180401802 on NCBI Build GRCm38
Downstream microRNA 133a-2 Mir133a-2 Mirn133a-2,
mir-133a-2
9.3kb Sense Chr. 2 - 180398379 - 180398482 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

regulation of gene expression
posttranscriptional regulation of gene expression
negative regulation of gene expression
negative regulation of translation
gene silencing by miRNA
regulation of calcineurin-NFAT signaling cascade

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10479375 - -
Affymetrix Mouse Clariom S Array PSR0200022815.mm.1 NR_029528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022815.mm.1 NR_029528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022815.mm.1 TC0200002790.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header