This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04279073_s1
See other 2410114N07RIK GE Assays ›
Gene Symbol:
2410114N07Rik
Gene Name:
RIKEN cDNA 2410114N07 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 34909789 - 34911481 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04279073_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2410114N07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040652.1 3 320 94
GenBank mRNA AK010767.1 - 3 321 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY710199.1 - 321 Chr. - - [-] on Build 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glycoprotein hormones,
alpha subunit
Cga CG-alpha,
FSHA,
GPHA1,
GPHalpha,
HCG,
LHA,
Tsha,
aGSU,
alpha-GSU,
alphaGSU,
alphaSU
-2.4kb Anti-sense Chr. 4 - 34893779 - 34907374 on NCBI Build GRCm38
Downstream MOB kinase activator 3B Mob3b 8430436F23Rik,
A430018A01Rik,
Mobkl2b
38.5kb Sense Chr. 4 - 34949074 - 35157495 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay targets the same location as MTA transcript cluster(s) TC0400002395.mm
This assay targets the same location as RTA transcript cluster(s) TC0500002502.rn

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945848 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793274 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36954 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128471 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03674217 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22833255 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22833260 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22833262 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22833259 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22833264 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400019458.mm.1 NR_040652.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019458.mm.1 NR_040652.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019458.mm.1 TC0400002395.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400019459.mm.1 NR_040652.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019459.mm.1 NR_040652.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019459.mm.1 TC0400002395.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400019457.mm.1 NR_040652.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019457.mm.1 NR_040652.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019457.mm.1 TC0400002395.mm.1 MTA TC
Murine Genome U74v2 Set 168597_at - -
Murine Genome U74v2 Set 168597_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 322002 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0