This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278829_m1
See other 4930452A19RIK GE Assays ›
Gene Symbol:
4930452A19Rik
Gene Name:
RIKEN cDNA 4930452A19 gene
Gene Aliases:
4933424P20Rik
Chromosome Location:
Chr.17: 9750752 - 9775866 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278829_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930452A19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045433.1 2 - 3 226 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream poly(A) binding protein,
cytoplasmic 6
Pabpc6 4932702K14Rik,
AI428050,
Pabpc3
-99.7kb Sense Chr. 17 - 9666497 - 9669704 on NCBI Build GRCm38
Downstream quaking Qk 1110003F05Rik,
1500005P18,
QkI,
l(17)-1Wis,
l17Wis1
427.8kb Sense Chr. 17 - 10202856 - 10319361 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header