This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278366_m1
See other 2010009K17RIK GE Assays ›
Gene Symbol:
2010009K17Rik
Gene Name:
RIKEN cDNA 2010009K17 gene
Gene Aliases:
-
Chromosome Location:
Chr.2: 158070364 - 158091797 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278366_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2010009K17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040609.1 2 - 3 246 73
GenBank mRNA AK008166.1 - 2 - 3 250 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY708124.1 - 2 - 3 249 Chr. 2 - 158070364 - 158091797 [-] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream regulation of nuclear pre-mRNA domain containing 1B Rprd1b 2610304G08Rik,
2810446G03Rik,
Crept
-5.9kb Anti-sense Chr. 2 - 158028489 - 158078207 on NCBI Build GRCm38
Downstream transglutaminase 2,
C polypeptide
Tgm2 G[a]h,
TG2,
TGase2,
tTG,
tTGas
29.8kb Sense Chr. 2 - 158116405 - 158146392 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005029.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200043042.mm.1 NR_040609.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043042.mm.1 NR_040609.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043042.mm.1 TC0200005029.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0