This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278220_m1
See other EPB4.1L4AOS GE Assays ›
Gene Symbol:
Epb4.1l4aos
Gene Name:
erythrocyte protein band 4.1 like 1 opposite strand
Gene Aliases:
2410004N09Rik
Chromosome Location:
Chr.18: 33794892 - 33795989 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278220_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1l4aos
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038152.1 1 - 2 236 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA W41357.1 - 183 Chr. - - [-] on Build 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 10549 Gm10549 ENSMUSG00000073610 -320.2kb Sense Chr. 18 - 33464163 - 33474710 on NCBI Build GRCm38
Downstream erythrocyte protein band 4.1 like 4a Epb4.1l4a Epb4.1l4,
Epb41l4a,
NBL4
338.0 Anti-sense Chr. 18 - 33796327 - 34007678 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000278.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800001989.mm.1 NR_038151.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001989.mm.1 NR_038151.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001989.mm.1 NR_038152.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001989.mm.1 NR_038152.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001989.mm.1 TC1800000278.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0