This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04242415_s1
See other SPEER5-PS1 GE Assays ›
Gene Symbol:
Speer5-ps1
Gene Name:
spermatogenesis associated glutamate (E)-rich protein 5, pseudogene 1
Gene Aliases:
1700007C10Rik, SPEER-5, Speer5-ps
Chromosome Location:
Chr.10: 44170447 - 44219585 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04242415_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Speer5-ps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_001582.2 7 1310 128
NR_027506.1 4 848 128
GenBank mRNA BB679670.1 - 1 291 128
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV206592.1 - 1 187 Chr. 10 - 44219277 - 44219583 [+] on Build GRCm38 128
GenBankmRNA AV039433.2 - 1 161 Chr. 10 - 44219303 - 44219583 [+] on Build GRCm38 128
GenBankmRNA AV210376.1 - 1 186 Chr. 10 - 44219278 - 44219583 [+] on Build GRCm38 128

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream absent in melanoma 1 Aim1 AI462491,
AI463325
-70.1kb Anti-sense Chr. 10 - 43950307 - 44148924 on NCBI Build GRCm38
Downstream predicted gene,
40637
Gm40637 48.2kb Anti-sense Chr. 10 - 44267742 - 44268243 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000412.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033278 NR_001582.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033278 NR_027506.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033278 BB679670.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033278 DV646842.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184508 NR_001582.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184508 NR_027506.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184508 DV646842.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184508 NR_001582.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184508 NR_027506.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184508 DV646842.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132223 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774551 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727504 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166044 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376386 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207911 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852828 AK035267.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852828 AK145464.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972096 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61896 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724593 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210110 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102404 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149626 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152661 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141278 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033278 NR_001582.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033278 NR_027506.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033278 BB679670.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033278 DV646842.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132223 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07045218 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141278 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07045185 - -
Whole Mouse Genome G4122A A_51_P184508 NR_001582.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184508 NR_027506.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184508 DV646842.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184508 NR_001582.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184508 NR_027506.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184508 DV646842.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134313 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141278 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08291499 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104262 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08216440 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104225 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104260 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08626303 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08941992 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104223 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104211 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104259 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08712838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104250 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08216439 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104240 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104214 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104198 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104200 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104243 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104283 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104213 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08739994 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104277 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104209 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104246 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104256 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104231 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08788976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104265 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08904156 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104248 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104219 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104252 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08452229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08900100 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104199 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104228 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27104224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08615062 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08216441 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430008_PM_x_at - -
Mouse Genome 430 2.0 Array 1430008_PM_x_at - -
Mouse Genome 430 2.0 Array 1430008_x_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 70365_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 70365_at - -
Affymetrix Mouse Clariom S Array PSR1000002803.mm.1 NR_027506.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002803.mm.1 TC1000000412.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000002809.mm.1 NR_001582.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000002809.mm.1 NR_027506.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002809.mm.1 TC1000000412.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000002811.mm.1 NR_001582.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000002811.mm.1 NR_027506.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002811.mm.1 TC1000000412.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000002810.mm.1 NR_001582.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000002810.mm.1 NR_027506.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002810.mm.1 TC1000000412.mm.1 MTA TC
Mouse Expression Set 430 1430008_PM_x_at - -
Mouse Expression Set 430 1430008_x_at - -
Murine Genome U74v2 Set 168149_PM_r_at - -
Murine Genome U74v2 Set 168149_r_at - -
Affymetrix HT Mouse Genome 430B Array 1430008_x_at - -
Affymetrix HT Mouse Genome 430B Array 1430008_PM_x_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 812394 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2850364 - -
Illumina mouse-6 v1.0 expression beadchip 2850364 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002850364 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2748533 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2748533 - -
mouse-6 v1.1 (Illumina) scl38955.8.1_275-S - -
Illumina Sentrix mouse-6.v1 bead array 2850364 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl38955.8.1_275-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850364 - -
MouseRef-8 v2 Expression BeadChip ILMN_2748533 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748533 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0