This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240907_s1
See other MIR1969 GE Assays ›
Gene Symbol:
Mir1969
Gene Name:
microRNA 1969
Gene Aliases:
mir-1969, mmu-mir-1969
Chromosome Location:
Chr.8: 70925525 - 70925618 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240907_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1969
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035495.1 1 44 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microtubule-associated protein 1S Map1s 6430517J16Rik,
Bpy2ip1,
Map8,
Mtap1s,
VCY2IP1
Sense Chr. 8 - 70905957 - 70930112 on NCBI Build GRCm38
Downstream predicted gene 10654 Gm10654 EG665828,
ENSMUSG00000074252
4.6kb Sense Chr. 8 - 70930203 - 70933053 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay targets the same location as MTA transcript cluster(s) TC0800000774.mm
This assay targets the same location as RTA transcript cluster(s) TC1200000874.rn

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800005506.mm.1 NR_035495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800005506.mm.1 NR_035495.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800005506.mm.1 TC0800000774.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0